Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
110 ml, 215 mm, Natural
Catalog # HIRS7780104
Supplier:   HIRSCHMANN
Measuring scoops
Catalog # HIRS7780104
Supplier:  HIRSCHMANN
Measuring scoops
110 ml, 215 mm, Natural
Price
28,50 €
Log in for availability
1 * 12 items
Product Details & Documents
HDPE, natural colour.
Specifications
  • Capacity:
    110 ml
  • Colour:
    Natural
  • Length:
    215 mm
  • Seq:
    04