Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
1250 ml, 400 mm, Natural
Catalog # HIRS7780108
Supplier:   HIRSCHMANN
Measuring scoops
Catalog # HIRS7780108
Supplier:  HIRSCHMANN
Measuring scoops
1250 ml, 400 mm, Natural
Price
67,30 €
Log in for availability
1 * 6 items
Product Details & Documents
HDPE, natural colour.
Specifications
  • Capacity:
    1250 ml
  • Colour:
    Natural
  • Length:
    400 mm
  • Seq:
    08