ContactUsLinkComponent
Agilent J&W PoraBOND Q, 50 m x 0.32 mm id, 5 µm, GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWLCP7352PT
Supplier:  Agilent
Agilent J&W PoraBOND Q, 50 m x 0.32 mm id, 5 µm, GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWLCP7352PT
Supplier:  Agilent
Frequently Bought Together