5 ppt NaCl Calibration Solution 1 * 480 mL
Catalog # EUTEECNACL5PPT
Supplier: EUTECH
Frequently Bought Together
VWR®, Melting point standards
Supplier: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion