Ice Bucket with lid, Magic Touch 2, black, 2.5L, expanded urethane, lightweight, chemical-resistant, snug lid, built-in pour spout, fill line indicator, raised top edge, easy transport, textured exterior, non-slip finish, 10.56D x 7.06inches. 1 * 12 items
Catalog # 216-2008
Supplier: Bel-Art Products, a Part of SP
Frequently Bought Together