Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
ClO2- Chlorites 1000 mg/l in H2O for IC 1 * 100 mL
ACSDH028.01O.L1
:
CPACHEM
ClO2- Chlorites 1000 mg/l in H2O for IC 1 * 100 mL
ACSDH028.01O.L1
:
CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Fluoride 1,000 mg/L in Water standard for ion chromatography
: VWR Chemicals
pH buffers, ready to use
: WTW
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Chlorite 1,000 mg/L in NaOH solution standard for ion chromatography
: VWR Chemicals
Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
: CPACHEM
Dodecamethylpentasiloxane, Sigma-Aldrich®
: Merck