Orders LinkContactUsLinkComponent
Capillary tubes
80 mm, 0,80 mm, 0,64 mm
Catalog # HILG1412213
Supplier:   HILGENBERG
Capillary tubes
Catalog # HILG1412213
Supplier:  HILGENBERG
Capillary tubes
Capillary tubes
80 mm, 0,80 mm, 0,64 mm
Price
557,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Soda-lime glass.
  • Both ends open
Specifications
  • Ext.Ø:
    0,80 mm
  • Ø int.:
    0,64 mm
  • Length:
    80 mm
  • Seq:
    07