Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
100 ml, Green
Catalog # 231-0414
Supplier:   VITLAB
Measuring scoops
Catalog # 231-0414
Supplier:  VITLAB
Measuring scoops
Measuring scoops
100 ml, Green
Price
20,60 €
Log in for availability
1 * 12 items
Product Details & Documents
PP, coloured. With precision formed filling edge and comfortable, stable handle.
  • Also suitable as weighing scoops
  • Easily readable volume quantities visible on the upper side of the handle
  • Food-safe product according to EC Directive No. 10/2011
Specifications
  • Capacity:
    100 ml
  • Colour:
    Green