Orders LinkContactUsLinkComponent
Measuring scoops
100 ml, Red
  231-0409
 :   VITLAB
Measuring scoops
  231-0409
 :  VITLAB
Measuring scoops
Measuring scoops
100 ml, Red
20,60 €
1 * 12 items
PP, coloured. With precision formed filling edge and comfortable, stable handle.
  • Also suitable as weighing scoops
  • Easily readable volume quantities visible on the upper side of the handle
  • Food-safe product according to EC Directive No. 10/2011
  • Capacity:
    100 ml
  • Colour:
    Red