Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer PCR optimized DyNAzyme (1x buffer contains: 10mM Tris-HCl (pH 8,8 at 25C), 1,5mM MgCl2, 50mM KCl and 0,1 % Triton X-100), 4x 1,5ml of 10x buffer 1 * 1 KIT
Catalog # F-511L
Supplier:
FINNZYMES REAGENTS
Expand image
Buffer PCR optimized DyNAzyme (1x buffer contains: 10mM Tris-HCl (pH 8,8 at 25C), 1,5mM MgCl2, 50mM KCl and 0,1 % Triton X-100), 4x 1,5ml of 10x buffer 1 * 1 KIT
Catalog # F-511L
Supplier:
FINNZYMES REAGENTS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection