Cylindrical bend testers
Cylindrical bend tester (100 mm) with desk holder and 14 mandrels (Ø = 2 - 32 mm)
Catalog # SIMESP1820
Supplier: Industrial Physics
Jump to:
Product Details & Documents
These cylindrical bend testers are used to determine the elasticity, adhesion and elongation of paint on metal.
- Sturdy conception in anodised aluminium and stainless steel
- Ergonomic clamping device for test panels
- Large knob on bending arm for smooth bending
The test panel is bent over the cylindrical mandrel and then checked for damages.
: ISO 1519 (or ASTM D522)
: Supplied with a desktop holder and mandrels.
Specifications
- Description:Cylindrical bend tester (100 mm) with desk holder and 14 mandrels (Ø = 2 - 32 mm)
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 6 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 10 mm
Product Information
- Catalog #AvailabilityPrice
- Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holderSet of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holderSIMESP1835Log in for availability1 * 14 items - 549,00 €
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 2 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 3 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 4 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 5 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 12 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 8 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 13 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 16 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 19 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 20 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 25 mm
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 32 mm
Product Information
- Catalog #AvailabilityPrice
- Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holderSet of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holderSIMESP1833Log in for availability1 * 1 items - 555,00 €
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 3/4"
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 1/8"
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 1/4"
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 3/8"
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 1/2"
Product Information
- Catalog #AvailabilityPrice
- Mandrel, 5/8"