ContactUsLinkComponent
Ice pan, solid, one piece molded, expanded polyvinyl chloride, unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 1 l capacity, blue.. 1 * 1 items
Catalog # BCISBCS-211B
Supplier:  BIOCISION
Ice pan, solid, one piece molded, expanded polyvinyl chloride, unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 1 l capacity, blue.. 1 * 1 items
Catalog # BCISBCS-211B
Supplier:  BIOCISION
Frequently Bought Together