Gooseneck light guides
Double combi light guide, 3 arms, Ø 4,5 mm, two gooseneck arms with length 500 mm, one flexible arm with length 800 mm, KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED
Catalog # SCOC170402
Supplier: SCHOTT AG Lighting and imaging
Jump to:
Product Details & Documents
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.
Documents
KL Series Fiber Optic Illumination for Stereo Microscopy
Product Literature
SCOC154101
Images 360
TDS Gooseneck Light Guides
Datasheet
Specifications
- Description:Double combi light guide, 3 arms, Ø 4,5 mm, two gooseneck arms with length 500 mm, one flexible arm with length 800 mm
- Seq:315
- For:KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED