Orders LinkContactUsLinkComponent
VWR® Separating Funnels, Squibb
50 ml, 19/26 NS
Catalog # 532-0018
Supplier:   VWR Collection
VWR® Separating Funnels, Squibb
Catalog # 532-0018
Supplier:  VWR Collection
VWR® Separating Funnels, Squibb
VWR® Separating Funnels, Squibb
50 ml, 19/26 NS
Price
50,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate 3.3 glass with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml
  • Seq:
    01