ContactUsLinkComponent
5 COMPOSES DANS H2O Chlorides (Cl-) 25mg/l Nitrites (NO2-) 75mg/l Nitrates (NO3-) 50mg/l Phosphates (PO43-) 75mg/l Sulphates (SO42-) 100mg/l 1 * 250 mL
Catalog # CMPL968B.25.W.L25
5 COMPOSES DANS H2O Chlorides (Cl-) 25mg/l Nitrites (NO2-) 75mg/l Nitrates (NO3-) 50mg/l Phosphates (PO43-) 75mg/l Sulphates (SO42-) 100mg/l 1 * 250 mL
Catalog # CMPL968B.25.W.L25
Frequently Bought Together