ContactUsLinkComponent
5 COMPOSES DANS H2O Chlorides (Cl-) 40mg/l Nitrites (NO2-) 10mg/l Nitrates (NO3-) 40mg/l Phosphates (PO43-) 20mg/l Sulphates (SO42-) 100mg/l 1 * 500 mL
Catalog # CMPLD371.40.W.L5
5 COMPOSES DANS H2O Chlorides (Cl-) 40mg/l Nitrites (NO2-) 10mg/l Nitrates (NO3-) 40mg/l Phosphates (PO43-) 20mg/l Sulphates (SO42-) 100mg/l 1 * 500 mL
Catalog # CMPLD371.40.W.L5
Frequently Bought Together