ACQUITY UPLC BEH Amide VanGuard Pre-column, 130Å, 1.7 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186004799
Supplier: WATERS
ACQUITY UPLC BEH Amide VanGuard Pre-column, 130Å, 1.7 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186004799
Supplier: WATERS
Frequently Bought Together
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Safety canisters, electrically conducting
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection