ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, G6PDH (L. MESENTEROIDES), NADP ASSAYED, LYOPHILIZED. PHOSPHOHEXOSE ISOMERASE, PHOSPHOGLUCONATE DEHYDROGE NASE, ADENYLATE KINASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES &LE 0.02%, 0.003%, 0.002% AND 0.002% RESPEC 1 * 1.000 EU
Catalog # ABNOP5264
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, G6PDH (L. MESENTEROIDES), NADP ASSAYED, LYOPHILIZED. PHOSPHOHEXOSE ISOMERASE, PHOSPHOGLUCONATE DEHYDROGE NASE, ADENYLATE KINASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES &LE 0.02%, 0.003%, 0.002% AND 0.002% RESPEC 1 * 1.000 EU
Catalog # ABNOP5264
Supplier:  Abnova
Related Information
Frequently Bought Together