ContactUsLinkComponent
KIT, ANA PROFILE ELISA KIT, ANA PROFILE ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUALITATIVE DETERMINATION ANA AUTOANTIBOD IES DIRECTED AGAINST RNP-70, RNP/SM, SM, SS-A, SS-B, SCL-70, CENTROMER B AND JO-1 IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1079
Supplier:  Abnova
KIT, ANA PROFILE ELISA KIT, ANA PROFILE ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUALITATIVE DETERMINATION ANA AUTOANTIBOD IES DIRECTED AGAINST RNP-70, RNP/SM, SM, SS-A, SS-B, SCL-70, CENTROMER B AND JO-1 IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1079
Supplier:  Abnova
Related Information
Frequently Bought Together