ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, PFU DNA POLYMERASE, IN ADDITION TO 5'-3' DNA POLYMERASE ACTIVITY, IT ALSO POSSESSES 3'-5' EXONUC LEASE (PROOFREADING) ACTIVITY. PFU DNA POLYMERASE EXHIBITS THE LOWEST ERROR RATE OF ANY THERMOSTABLE DNA POLYMERASE S 1 * 500 EU
Catalog # ABNOD0014
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, PFU DNA POLYMERASE, IN ADDITION TO 5'-3' DNA POLYMERASE ACTIVITY, IT ALSO POSSESSES 3'-5' EXONUC LEASE (PROOFREADING) ACTIVITY. PFU DNA POLYMERASE EXHIBITS THE LOWEST ERROR RATE OF ANY THERMOSTABLE DNA POLYMERASE S 1 * 500 EU
Catalog # ABNOD0014
Supplier:  Abnova
Frequently Bought Together