ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, TAQ DNA POLYMERASE, TAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PRO VED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABLE AT H 1 * 500 EU
Catalog # ABNOD0010
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, TAQ DNA POLYMERASE, TAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PRO VED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABLE AT H 1 * 500 EU
Catalog # ABNOD0010
Supplier:  Abnova
Frequently Bought Together