Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7 (1 mol/L)
TRIS buffer solution pH 7 (1 mol/L), Glass bottle, 500 mL
Catalog # CAYM600200-500
Supplier:   Cayman Chemical
Buffer, TRIS solution pH 7 (1 mol/L)
Catalog # CAYM600200-500
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TRIS solution pH 7 (1 mol/L)
TRIS buffer solution pH 7 (1 mol/L), Glass bottle, 500 mL
CAS Number:  
Price
121,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TRIS buffer solution pH 7 (1 mol/L)