3-Aminothiobenzamide
Glass bottle for solids, 500 mg
Catalog # CAYM11064-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3-Aminobenzothioamide
- CAS Number:78950-36-4
- Formula:C₇H₈N₂S
- MDL Number:MFCD04973325
- Molecular Weight:152.22 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



