Orders LinkContactUsLinkComponent
Estrone
Estrone
Glass bottle for solids, 10 g
Catalog # CAYM10006485-10
Supplier:   Cayman Chemical
Estrone
Catalog # CAYM10006485-10
Supplier:  Cayman Chemical
CAS Number:  53-16-7
Estrone
Glass bottle for solids, 10 g
CAS Number:  53-16-7
Price
195,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • MDL Number:MFCD00003620
  • EINECS:200-164-5
Documents
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
Specifications