Orders LinkContactUsLinkComponent
Needles, Spinocan®
Needles, Spinocan®
Needles, Spinocan®, 19, 88 mm, 1,10 mm, Beige
Catalog # BRAU4501195-13
Supplier:   B. Braun
Needles, Spinocan®
Catalog # BRAU4501195-13
Supplier:  B. Braun
Needles, Spinocan®
Needles, Spinocan®
Needles, Spinocan®, 19, 88 mm, 1,10 mm, Beige
Price
139,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Specifications
  • Colour code:
    Beige
  • Ø:
    1,10 mm
  • Gauge:
    19
  • Description:
    Needles, Spinocan®
  • Length:
    88 mm
  • Seq:
    004