ContactUsLinkComponent
Eclipse Ci-S Main Body including Capture Button, Fixed Diffuser, Field Diaphragm, Adjusting Tool, Dual Coarse/Fine Focusing with refocusing mechanism and torque adjustment, and Built-in ND4, ND8, NCB-11 Filters, 6V 30W Lamphouse & Vinyl Cover 104C 1 * 1 items
Catalog # NIKOMBA94010
Supplier:  NIKON
Eclipse Ci-S Main Body including Capture Button, Fixed Diffuser, Field Diaphragm, Adjusting Tool, Dual Coarse/Fine Focusing with refocusing mechanism and torque adjustment, and Built-in ND4, ND8, NCB-11 Filters, 6V 30W Lamphouse & Vinyl Cover 104C 1 * 1 items
Catalog # NIKOMBA94010
Supplier:  NIKON
Frequently Bought Together