Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade
Agar, Bacteriological Grade
Bottle, 1 kg
Catalog # RC-004
Supplier:   G-Biosciences
Agar, Bacteriological Grade
Catalog # RC-004
Supplier:  G-Biosciences
CAS Number:  9002-18-0
Agar, Bacteriological Grade
Agar, Bacteriological Grade
Bottle, 1 kg
CAS Number:  9002-18-0
Price
418,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.
Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:(C₁₂H₁₈O₉)n
  • MDL Number:MFCD00081288
  • Melting Pt:90 °C
Specifications
Specifications
  • Pack type:
    Bottle
  • Pk:
    1 kg
Specifications