Ready-to-use 1 mL and 5 mL pre-packed PRC columns.Q and S HyperCel sorbents are composed of a rigid cellulose matrix with excellent flow properties and low backpressure, providing high dynamic binding capacity at short residence time. PRC COLUMN 8X10 1 * 1 items
Catalog # 518-0085
Supplier: Cytiva (Formerly Pall Lab)
Frequently Bought Together