ContactUsLinkComponent
Ready-to-use 1 mL and 5 mL pre-packed PRC columns allow direct connection to chromatography systems. Q and CM Ceramic HyperD sorbent s allow for high dynamic binding capacity independent of flow rate. 1 * 1 items
  518-0076
Ready-to-use 1 mL and 5 mL pre-packed PRC columns allow direct connection to chromatography systems. Q and CM Ceramic HyperD sorbent s allow for high dynamic binding capacity independent of flow rate. 1 * 1 items
  518-0076
Frequently Bought Together