ContactUsLinkComponent
Pipet Tip 300 µL Pre-Sterile, Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: FlexFit provides universal fit and superior seal, X-Resin for low sample retention, Blade for accuracy, serial number for the best tra 1 * 960 items
Catalog # 613-1779
Supplier:  BIOTIX
Pipet Tip 300 µL Pre-Sterile, Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: FlexFit provides universal fit and superior seal, X-Resin for low sample retention, Blade for accuracy, serial number for the best tra 1 * 960 items
Catalog # 613-1779
Supplier:  BIOTIX
Frequently Bought Together