ContactUsLinkComponent
Pipet Tip 10XL Pre-Sterile, Tip Eject Reload System 96 Tip/Card, 20 Cards/Pack, 5 Pack/Case. Patented features: FlexFit provides un iversal fit and superior seal, X-Resin for low sample retention, Blade for accuracy, serial number for the best tracea 1 * 1.920 items
Catalog # 613-1776
Supplier:  BIOTIX
Pipet Tip 10XL Pre-Sterile, Tip Eject Reload System 96 Tip/Card, 20 Cards/Pack, 5 Pack/Case. Patented features: FlexFit provides un iversal fit and superior seal, X-Resin for low sample retention, Blade for accuracy, serial number for the best tracea 1 * 1.920 items
Catalog # 613-1776
Supplier:  BIOTIX
Frequently Bought Together