Tubes and systems for active sampling
Activated charcoal tubes, G
Catalog # DRAE6400745
Supplier: Dräger
Jump to:
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Specifications
- Type:G
- Description:Activated charcoal tubes
- Seq:08