Sartobind capsules are especially suited for the removal of DNA, viruses and protein contaminants in biopharmaceutical production. S artobind membranes contain homogenelously grafted binding sites on a reinforced cross-linked cellulose matrix. The la 1 * 4 items
Catalog # SART92IEXS42DN-11A
Supplier: Sartorius
Frequently Bought Together