ContactUsLinkComponent
The Vivacon 500 PCR Grade units have been treated with a dual-cycle ethylene oxide (ETO) gas process to ensure they are sterile to a safety assurance level of 106 and free of detectable human genomic DNA contamination. Tests were performed with a qP 1 * 100 items
Catalog # 518-0062
Supplier:  Sartorius
The Vivacon 500 PCR Grade units have been treated with a dual-cycle ethylene oxide (ETO) gas process to ensure they are sterile to a safety assurance level of 106 and free of detectable human genomic DNA contamination. Tests were performed with a qP 1 * 100 items
Catalog # 518-0062
Supplier:  Sartorius
Frequently Bought Together