ContactUsLinkComponent
13 COMPOSES DANS HNO3 5% Ca 10000mg/l Mg 10000mg/l K 10000mg/l Na 10000mg/l P 500mg/l Mn 50mg/l Fe 50mg/l Cr 50mg/l Al 50mg/l Cu 50mg/l Ni 50mg/l B 100mg/l Zn 100mg/l 1 * 100 mL
  CMPL24A4.50.5N.L1
13 COMPOSES DANS HNO3 5% Ca 10000mg/l Mg 10000mg/l K 10000mg/l Na 10000mg/l P 500mg/l Mn 50mg/l Fe 50mg/l Cr 50mg/l Al 50mg/l Cu 50mg/l Ni 50mg/l B 100mg/l Zn 100mg/l 1 * 100 mL
  CMPL24A4.50.5N.L1
Frequently Bought Together