Orders LinkContactUsLinkComponent
Scoops
Scoops
600 ml, 300 mm, HDPE, White
Catalog # LAMA64
Supplier:   LAMAPLAST
Scoops
Catalog # LAMA64
Supplier:  LAMAPLAST
Scoops
Scoops
600 ml, 300 mm, HDPE, White
Price
6,10 €
Log in for availability
1 * 1 items
Product Details & Documents
These scoops are made of high density PE. Ideal for use in food applications.
  • For withdrawal of solid and granulates samples
  • Rounded bottom
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Specifications
  • Capacity:
    600 ml
  • Colour:
    White
  • Length:
    300 mm
  • Material:
    HDPE
  • Seq:
    03