Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
130 mm, 4 mm
Catalog # 231-0464
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0464
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
130 mm, 4 mm
Price
36,50 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades
Documents
Specifications
  • Length:
    130 mm
  • Blade width:
    4 mm