Beaker cups, medicine
30 ml, Red
Catalog # HERE1090509R
Supplier: HERENZ HEINZ
Jump to:
Product Details & Documents
PP, translucent.
- Graduated medicinal cups
- Available in different colours
- Unbreakable
Documents
Specifications
- Capacity:30 ml
- Colour:Red
- Seq:04