Orders LinkContactUsLinkComponent
Beaker cups, medicine
Beaker cups, medicine
30 ml, Red
Catalog # HERE1090509R
Supplier:   HERENZ HEINZ
Beaker cups, medicine
Catalog # HERE1090509R
Supplier:  HERENZ HEINZ
Beaker cups, medicine
Beaker cups, medicine
30 ml, Red
Price
Inquire For Price
Log in for availability
1 * 3.750 items
Product Details & Documents
PP, translucent.
  • Graduated medicinal cups
  • Available in different colours
  • Unbreakable
Specifications
  • Capacity:
    30 ml
  • Colour:
    Red
  • Seq:
    04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.