Coverall flameresistant ProShield® FR - white with orange seams, model CHF5, Size L. 3-piece hood, external stitched orange colour-coded seams, zipper flap, elasticated waist, elasticated cuffs, ankles & hood, antistatic treatment on both sides. 1 * 100 items
Catalog # 113-6385
Supplier: DuPont
Frequently Bought Together