SOLUTION MULTIELEMENTAIRE DE 12 COMPOSES DANS HNO3 5% Cd 500mg/l Ca 1000mg/l Cr 1000mg/l Fe 1000mg/l Ni 1000mg/l Pb 2500mg/l Zn 2500mg/l Se 50mg/l Cu 50mg/l K 50mg/l P 50mg/l Ba 1000mg/l 1 * 250 mL
Catalog # CMPL9DD0.1K.5N.L25
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together