Needles, Spinocan®
Needles, Spinocan®, 22, 88 mm, 0,70 mm, Black
Catalog # BRAU4507908
Supplier: B. Braun
Jump to:
Product Details & Documents
Specifications
- Colour code:Black
- Ø:0,70 mm
- Gauge:22
- Description:Needles, Spinocan®
- Length:88 mm
- Seq:008