Orders LinkContactUsLinkComponent
Lab markers
LabMarker, fine tip, Black
Catalog # 811-0177
Supplier:   MICRONOVA
Lab markers
Catalog # 811-0177
Supplier:  MICRONOVA
Lab markers
Lab markers
LabMarker, fine tip, Black
Price
98,10 €
Log in for availability
1 * 10 items
Product Details & Documents
LabMarkers feature permanent ink that marks on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal or porcelain labware but can be scrubbed off.
  • Fine tip, black ink
  • Permanent on most surfaces
  • Fast drying
  • Available gamma irradiated
Materials of construction: High-density poly barrel. Permanent, quick drying, alcohol- resistant black ink.
Chemical compatibility: Holds up to wipedown with IPA, bleach, and phenols. Integrity of printed barrel will be compromised with thinners and strong solvents.
 : Markers cannot be used with Ethanol or other alcohols.
Specifications
  • Colour:
    Black
  • Description:
    LabMarker, fine tip
  • Seq:
    01