5 COMPOSES POUR IC DANS H2O Nitrites (NO2-) 0.04mg/l Nitrates (NO3-) 0.4mg/l Chlorides (Cl-) 40mg/l Ammonium (NH4+) 0.4mg/l Phosphates (PO43-) 0.03mg/l 1 * 100 mL
Catalog # CMPL54F1.40.W.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together