Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Gilson type, 200 - 1000 µl, Transparent, Bulk, 500/bag
Catalog # LPIT152146
Supplier:   LP ITALIANA
Pipette tips
Catalog # LPIT152146
Supplier:  LP ITALIANA
Pipette tips
Pipette tips
Gilson type, 200 - 1000 µl, Transparent, Bulk, 500/bag
Price
228,00 €
Log in for availability
1 * 11.000 items
Product Details & Documents
Specifications
  • Packed:
    Bulk, 500/bag
  • Volume:
    200 - 1000 µl
  • Tip style:
    Gilson type
  • Tip colour:
    Transparent