ContactUsLinkComponent
Refrigerated bath/circulator, optima tx150 + r5 refrigeration unit, 12l capacity, incl. drain tap. temp range -47 to 100C. 1 * 1 items
Catalog # 461-0705
Supplier:  GRANT INSTRUMENTS
Refrigerated bath/circulator, optima tx150 + r5 refrigeration unit, 12l capacity, incl. drain tap. temp range -47 to 100C. 1 * 1 items
Catalog # 461-0705
Supplier:  GRANT INSTRUMENTS
Related Information
Frequently Bought Together