ContactUsLinkComponent
Refrigerated bath/circulator, optima tx150 + r2 refrigeration unit, 5l capacity, incl. drain tap. temp range -20 to 100C. 1 * 1 items
Catalog # 461-0702
Supplier:  GRANT INSTRUMENTS
Refrigerated bath/circulator, optima tx150 + r2 refrigeration unit, 5l capacity, incl. drain tap. temp range -20 to 100C. 1 * 1 items
Catalog # 461-0702
Supplier:  GRANT INSTRUMENTS