ContactUsLinkComponent
Refrigerated bath, optima t100 + r4 refrigeration unit, 20l capacity, incl. drain tap, temp range 0 to 100C. 1 * 1 items
Catalog # 461-0685
Supplier:  GRANT INSTRUMENTS
Refrigerated bath, optima t100 + r4 refrigeration unit, 20l capacity, incl. drain tap, temp range 0 to 100C. 1 * 1 items
Catalog # 461-0685
Supplier:  GRANT INSTRUMENTS
Related Information
Frequently Bought Together