STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST18 STAINLESS STEEL TANK, 18L CAPACITY, INCL DRAIN TAP AN D PUMP. TEMP RANGE AMBIENT +5 TO 150C. 1 * 1 items
Catalog # GRANTX150-ST18EU
Supplier: GRANT INSTRUMENTS
Frequently Bought Together