ContactUsLinkComponent
Optima txf200 immersion thermostat, integrated variable speed pump for external circulation, programmable. temp range amb. +5 to 2 00C. 1 * 1 items
Catalog # GRANTXF200EU
Supplier:  GRANT INSTRUMENTS
Optima txf200 immersion thermostat, integrated variable speed pump for external circulation, programmable. temp range amb. +5 to 2 00C. 1 * 1 items
Catalog # GRANTXF200EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together