STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT+5 TO 200C. 1 * 1 items
Catalog # GRANTXF200-ST26EU
Supplier: GRANT INSTRUMENTS
Frequently Bought Together